Pyridine-[D₅] ≥99%

Supplier: Matrix Scientific

Synonyms: Pyridine-[²H₅]

086136-1G
102847-066EA 127.69 USD
102847-066
Pyridine-[D₅] ≥99%
Pyridine-[D5]

Hazard: Irritant

Formula: C₅D₅N
MW: 84.06 g/mol
MDL Number: MFCD00044639
CAS Number: 7291-22-7
UN: 1282
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR