2-Ethyl-2-methyl-1,2,3-trihydroquinazolin-4-one ≥95%

Supplier: Matrix Scientific

086015-1G
102846-646EA 666.15 USD
102846-646
2-Ethyl-2-methyl-1,2,3-trihydroquinazolin-4-one ≥95%
2-Ethyl-2-methyl-1,2,3-trihydroquinazolin-4-one

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MW: 190.25 g/mol
MDL Number: MFCD00171144
CAS Number: 1012-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR