5-(Trimethylsilylethynyl)indane ≥97%

Supplier: Matrix Scientific

086000-1G
102846-610EA 553.85 USD
102846-610
5-(Trimethylsilylethynyl)indane ≥97%
5-(Trimethylsilylethynyl)indane

Hazard: Irritant

Formula: C₁₄H₁₈Si
MDL Number: MFCD09991849
CAS Number: 1216812-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR