N-2,3-Dihydro-1H-inden-1-ylmethanesulfonamide ≥97%

Supplier: Matrix Scientific

085998-1G
102846-606EA 340 USD
102846-606
N-2,3-Dihydro-1H-inden-1-ylmethanesulfonamide ≥97%
N-2,3-Dihydro-1H-inden-1-ylmethanesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₃NO₂S
MDL Number: MFCD09991847
CAS Number: 294209-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR