3-(4-Aminobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%

Supplier: Matrix Scientific

085997-1G
102846-604EA 426.15 USD
102846-604
3-(4-Aminobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol ≥95%
3-(4-Aminobenzyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-ol

Hazard: Irritant

Formula: C₁₁H₁₀N₆O
MDL Number: MFCD09864986
CAS Number: 1216756-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR