6-(Piperazin-1-ylcarbonyl)-2-naphthol ≥97%

Supplier: Matrix Scientific

085935-1G
102846-426EA 806.15 USD
102846-426
6-(Piperazin-1-ylcarbonyl)-2-naphthol ≥97%
6-(Piperazin-1-ylcarbonyl)-2-naphthol

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD18089283
CAS Number: 1287217-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR