tert-Butyl (2S)-2-isopropyl-4-oxopyrrolidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

085784-1G
102845-980EA 1344.62 USD
102845-980
tert-Butyl (2S)-2-isopropyl-4-oxopyrrolidine-1-carboxylate ≥97%
tert-Butyl (2S)-2-isopropyl-4-oxopyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁NO₃
MDL Number: MFCD08056351
CAS Number: 1212437-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR