tert-Butyl-4-tetrahydro-2H-pyran-4-ylpiperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(tetrahydro-2H-pyran-4-yl)piperazine-1-carboxylate

085670-1G
102845-650EA 723.08 USD
102845-650
tert-Butyl-4-tetrahydro-2H-pyran-4-ylpiperazine-1-carboxylate ≥97%
tert-Butyl-4-tetrahydro-2H-pyran-4-ylpiperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₃
MW: 270.37 g/mol
MDL Number: MFCD11505064
CAS Number: 706759-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR