tert-Butyl-4-[(4S)-4-benzyl-2-oxo-1,3-oxazolidin-3-yl]piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

085625-1G
102845-528EA 1032.31 USD
102845-528
tert-Butyl-4-[(4S)-4-benzyl-2-oxo-1,3-oxazolidin-3-yl]piperidine-1-carboxylate ≥97%
tert-Butyl-4-[(4S)-4-benzyl-2-oxo-1,3-oxazolidin-3-yl]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₀H₂₈N₂O₄
MW: 360.46 g/mol
MDL Number: MFCD09991845
CAS Number: 1217482-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR