tert-Butyl-4-(5,5-dimethyl-2-oxo-1,3-oxazinan-3-yl)piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

085617-1G
102845-512EA 723.08 USD
102845-512
tert-Butyl-4-(5,5-dimethyl-2-oxo-1,3-oxazinan-3-yl)piperidine-1-carboxylate ≥97%
tert-Butyl-4-(5,5-dimethyl-2-oxo-1,3-oxazinan-3-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₈N₂O₄
MW: 312.41 g/mol
MDL Number: MFCD09991837
CAS Number: 1215596-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR