tert-Butyl-4-oxooctahydroquinoline-1(2H)-carboxylate ≥97%

Supplier: Matrix Scientific

085560-1G
102845-368EA 949.23 USD
102845-368
tert-Butyl-4-oxooctahydroquinoline-1(2H)-carboxylate ≥97%
tert-Butyl-4-oxooctahydroquinoline-1(2H)-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₃NO₃
MW: 253.34 g/mol
MDL Number: MFCD18089270
CAS Number: 1287218-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR