4,5-Methyl-1,3-dihydro-2H-benzimidazol-2-one ≥95%

Supplier: Matrix Scientific

085546-1G
102845-324EA 353.85 USD
102845-324
4,5-Methyl-1,3-dihydro-2H-benzimidazol-2-one ≥95%
4,5-Methyl-1,3-dihydro-2H-benzimidazol-2-one

Hazard: Irritant

Formula: C₉H₁₀N₂O
MDL Number: MFCD08274660
CAS Number: 1217032-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR