4-Methyl-1,3-benzoxazol-2(3H)-one ≥97%

Supplier: Matrix Scientific

085536-1G
102845-298EA 949.23 USD
102845-298
4-Methyl-1,3-benzoxazol-2(3H)-one ≥97%
4-Methyl-1,3-benzoxazol-2(3H)-one

Hazard: Irritant

Formula: C₈H₇NO₂
MDL Number: MFCD16742805
CAS Number: 78258-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR