5-(7-Ethoxy-1-benzofuran-2-yl)-1,3,4-oxadiazol-2(3H)-one ≥97%

Supplier: Matrix Scientific

085505-1G
102845-214EA 524.62 USD
102845-214
5-(7-Ethoxy-1-benzofuran-2-yl)-1,3,4-oxadiazol-2(3H)-one ≥97%
5-(7-Ethoxy-1-benzofuran-2-yl)-1,3,4-oxadiazol-2(3H)-one

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄
MDL Number: MFCD09991830
CAS Number: 1216537-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR