4-[6-(Trifluoromethyl)pyridazin-3-yl]morpholine ≥97%

Supplier: Matrix Scientific

085470-1G
102845-120EA 806.15 USD
102845-120
4-[6-(Trifluoromethyl)pyridazin-3-yl]morpholine ≥97%
4-[6-(Trifluoromethyl)pyridazin-3-yl]morpholine

Hazard: Irritant

Formula: C₉H₁₀F₃N₃O
MDL Number: MFCD11840305
CAS Number: 1215530-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR