N-[(Dimethylamino)methylene]pyrazine-2-carboxamide ≥97%

Supplier: Matrix Scientific

085430-1G
102845-034EA 383.08 USD
102845-034
N-[(Dimethylamino)methylene]pyrazine-2-carboxamide ≥97%
N-[(Dimethylamino)methylene]pyrazine-2-carboxamide

Hazard: Irritant

Formula: C₈H₁₀N₄O
MDL Number: MFCD11100141
CAS Number: 78132-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR