2-Hydrazino-5-methyl-4,5-dihydro-1H-imidazole hydroiodide ≥97%

Supplier: Matrix Scientific

085392-1G
102844-926EA 481.54 USD
102844-926
2-Hydrazino-5-methyl-4,5-dihydro-1H-imidazole hydroiodide ≥97%
2-Hydrazino-5-methyl-4,5-dihydro-1H-imidazole hydroiodide

Hazard: Irritant

Formula: C₄H₁₁IN₄
MDL Number: MFCD09258877
CAS Number: 49541-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR