1-[(5-Chloro-1-methyl-1H-pyrazol-3-yl)carbonyl]piperidine ≥97%

Supplier: Matrix Scientific

085365-1G
102844-860EA 949.23 USD
102844-860
1-[(5-Chloro-1-methyl-1H-pyrazol-3-yl)carbonyl]piperidine ≥97%
1-[(5-Chloro-1-methyl-1H-pyrazol-3-yl)carbonyl]piperidine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃O
MDL Number: MFCD00106216
CAS Number: 648408-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR