2-{[(2-Acetylthien-3-yl)amino]carbonyl}benzoic acid ≥97%

Supplier: Matrix Scientific

085270-1G
102844-620EA 806.15 USD
102844-620
2-{[(2-Acetylthien-3-yl)amino]carbonyl}benzoic acid ≥97%
2-{[(2-Acetylthien-3-yl)amino]carbonyl}benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁NO₄S
MDL Number: MFCD00102651
CAS Number: 175602-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR