N-(1-Methylpiperidin-4-yl)-3-nitropyridin-2-amine ≥97%

Supplier: Matrix Scientific

085207-1G
102844-458EA 949.23 USD
102844-458
N-(1-Methylpiperidin-4-yl)-3-nitropyridin-2-amine ≥97%
N-(1-Methylpiperidin-4-yl)-3-nitropyridin-2-amine

Hazard: Irritant

Formula: C₁₁H₁₆N₄O₂
MDL Number: MFCD12408759
CAS Number: 1096347-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR