N-Benzyl-N-isopropyl-3-nitropyridin-2-amine ≥97%
Supplier: Matrix Scientific
085193-1G
102844-430EA
863.08
USD
102844-430
N-Benzyl-N-isopropyl-3-nitropyridin-2-amine ≥97%
N-Benzyl-N-isopropyl-3-nitropyridin-2-amine
Hazard: Irritant
Formula:
C₁₅H₁₇N₃O₂ |
MDL Number:
MFCD21099395 CAS Number: 1353953-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...