N-(2-Methoxy-1-methylethyl)-3-nitropyridin-2-amine ≥97%

Supplier: Matrix Scientific

085174-1G
102844-386EA 949.23 USD
102844-386
N-(2-Methoxy-1-methylethyl)-3-nitropyridin-2-amine ≥97%
N-(2-Methoxy-1-methylethyl)-3-nitropyridin-2-amine

Hazard: Irritant

Formula: C₉H₁₃N₃O₃
MDL Number: MFCD11115137
CAS Number: 1036536-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR