3-Nitro-2-phenoxypyridine ≥97%

Supplier: Matrix Scientific

085140-1G
102844-314EA 467.69 USD
102844-314
3-Nitro-2-phenoxypyridine ≥97%
3-Nitro-2-phenoxypyridine

Hazard: Irritant

Formula: C₁₁H₈N₂O₃
MDL Number: MFCD01928026
CAS Number: 76893-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR