6-(Tetrahydro-2H-pyran-2-ylmethoxy)nicotinonitrile ≥97%

Supplier: Matrix Scientific

085125-1G
102844-278EA 553.85 USD
102844-278
6-(Tetrahydro-2H-pyran-2-ylmethoxy)nicotinonitrile ≥97%
6-(Tetrahydro-2H-pyran-2-ylmethoxy)nicotinonitrile

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD09942913
CAS Number: 1016889-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR