2-(Tetrahydro-2H-pyran-2-ylmethoxy)isonicotinonitrile ≥97%

Supplier: Matrix Scientific

085124-1G
102844-276EA 553.85 USD
102844-276
2-(Tetrahydro-2H-pyran-2-ylmethoxy)isonicotinonitrile ≥97%
2-(Tetrahydro-2H-pyran-2-ylmethoxy)isonicotinonitrile

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
MW: 218.26 g/mol
MDL Number: MFCD09950970
CAS Number: 1016791-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR