2-(Cyclobutylmethoxy)nicotinonitrile ≥97%

Supplier: Matrix Scientific

085122-1G
102844-272EA 553.85 USD
102844-272
2-(Cyclobutylmethoxy)nicotinonitrile ≥97%
2-(Cyclobutylmethoxy)nicotinonitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD13195631
CAS Number: 1235440-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR