2-(Cyclobutylmethoxy)isonicotinonitrile ≥97%

Supplier: Matrix Scientific

085116-1G
102844-260EA 553.85 USD
102844-260
2-(Cyclobutylmethoxy)isonicotinonitrile ≥97%
2-(Cyclobutylmethoxy)isonicotinonitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD13195627
CAS Number: 1235440-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR