2-(2-Furyl)isonicotinonitrile ≥97%

Supplier: Matrix Scientific

085114-1G
102844-254EA 553.85 USD
102844-254
2-(2-Furyl)isonicotinonitrile ≥97%
2-(2-Furyl)isonicotinonitrile

Hazard: Irritant

Formula: C₁₀H₆N₂O
MDL Number: MFCD13195626
CAS Number: 1235440-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR