1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone ≥95%
Supplier: Matrix Scientific
085090-1G
102844-200EA
469.23
USD
102844-200
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone ≥95%
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone
Hazard: Irritant
Formula:
C₁₄H₁₃NO₂ |
MDL Number:
MFCD12404914 CAS Number: 1215576-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...