1-[6-(3-Chlorophenyl)pyridin-3-yl]ethanone ≥97%

Supplier: Matrix Scientific

085089-1G
102844-198EA 503.08 USD
102844-198
1-[6-(3-Chlorophenyl)pyridin-3-yl]ethanone ≥97%
1-[6-(3-Chlorophenyl)pyridin-3-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₀ClNO
MDL Number: MFCD12404913
CAS Number: 1216636-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR