1-[6-(4-Chlorophenyl)pyridin-3-yl]ethanone ≥97%

Supplier: Matrix Scientific

085088-1G
102844-196EA 723.08 USD
102844-196
1-[6-(4-Chlorophenyl)pyridin-3-yl]ethanone ≥97%
1-[6-(4-Chlorophenyl)pyridin-3-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₀ClNO
MDL Number: MFCD12404912
CAS Number: 1216474-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR