2-Chloro-4-pyridin-3-yl-1,3,5-triazine ≥97%

Supplier: Matrix Scientific

084066-1G
102841-432EA 750.77 USD
102841-432
2-Chloro-4-pyridin-3-yl-1,3,5-triazine ≥97%
2-Chloro-4-pyridin-3-yl-1,3,5-triazine

Hazard: Irritant

Formula: C₈H₅ClN₄
MDL Number: MFCD09864962
CAS Number: 1053656-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR