3-Bromo-1-(1,3-oxazol-2-yl)pyrrolidin-2-one ≥97%
Supplier: Matrix Scientific
084060-1G
102841-422EA
1430.77
USD
102841-422
3-Bromo-1-(1,3-oxazol-2-yl)pyrrolidin-2-one ≥97%
3-Bromo-1-(1,3-oxazol-2-yl)pyrrolidin-2-one
Hazard: Irritant
Formula:
C₇H₇BrN₂O₂ |
MDL Number:
MFCD18089251 CAS Number: 1287217-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...