3-Bromo-1-pyridin-3-ylpyrrolidin-2-one ≥97%

Supplier: Matrix Scientific

084055-1G
102841-412EA 1107.69 USD
102841-412
3-Bromo-1-pyridin-3-ylpyrrolidin-2-one ≥97%
3-Bromo-1-pyridin-3-ylpyrrolidin-2-one

Hazard: Irritant

Formula: C₉H₉BrN₂O
MDL Number: MFCD18089246
CAS Number: 178946-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR