5-Bromo-7-(trifluoromethyl)quinoxaline ≥97%

Supplier: Matrix Scientific

084043-1G
102841-380EA 524.62 USD
102841-380
5-Bromo-7-(trifluoromethyl)quinoxaline ≥97%
5-Bromo-7-(trifluoromethyl)quinoxaline

Hazard: Irritant

Formula: C₉H₄BrF₃N₂
MDL Number: MFCD18089243
CAS Number: 1287218-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR