3-Chloro-6-(tetrahydro-2H-pyran-2-ylmethoxy)pyridazine ≥97%
Supplier: Matrix Scientific
083997-1G
102841-264EA
750.77
USD
102841-264
3-Chloro-6-(tetrahydro-2H-pyran-2-ylmethoxy)pyridazine ≥97%
3-Chloro-6-(tetrahydro-2H-pyran-2-ylmethoxy)pyridazine
Hazard: Irritant
Formula:
C₁₀H₁₃ClN₂O₂ |
MDL Number:
MFCD09809808 CAS Number: 1016697-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...