3-Chloro-6-(cyclobutylmethoxy)pyridazine ≥97%
Supplier: Matrix Scientific
083996-1G
102841-262EA
636.92
USD
102841-262
3-Chloro-6-(cyclobutylmethoxy)pyridazine ≥97%
3-Chloro-6-(cyclobutylmethoxy)pyridazine
Hazard: Irritant
Formula:
C₉H₁₁ClN₂O |
MDL Number:
MFCD14666501 CAS Number: 1287217-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...