3-Chloro-6-(tetrahydro-2H-pyran-4-yloxy)pyridazine ≥97%
Supplier: Matrix Scientific
083995-1G
102841-260EA
750.77
USD
102841-260
3-Chloro-6-(tetrahydro-2H-pyran-4-yloxy)pyridazine ≥97%
3-Chloro-6-(tetrahydro-2H-pyran-4-yloxy)pyridazine
Hazard: Irritant
Formula:
C₉H₁₁ClN₂O₂ |
MDL Number:
MFCD14614943 CAS Number: 1251393-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...