2-Chloro-4-phenoxypyrimidine ≥97%

Supplier: Matrix Scientific

083987-1G
102841-238EA 750.77 USD
102841-238
2-Chloro-4-phenoxypyrimidine ≥97%
2-Chloro-4-phenoxypyrimidine

Hazard: Irritant

Formula: C₁₀H₇ClN₂O
MDL Number: MFCD16662235
CAS Number: 18214-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR