4-Chloro-6-methyl-2-(1H-pyrrol-1-yl)pyrimidine ≥97%

Supplier: Matrix Scientific

083986-1G
102841-236EA 750.77 USD
102841-236
4-Chloro-6-methyl-2-(1H-pyrrol-1-yl)pyrimidine ≥97%
4-Chloro-6-methyl-2-(1H-pyrrol-1-yl)pyrimidine

Hazard: Irritant

Formula: C₉H₈ClN₃
MDL Number: MFCD19706040
CAS Number: 1355334-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR