2-Chloro-N-(3-bromophenyl)pyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

083976-1G
102841-212EA 467.69 USD
102841-212
2-Chloro-N-(3-bromophenyl)pyrimidin-4-amine ≥97%
2-Chloro-N-(3-bromophenyl)pyrimidin-4-amine

Hazard: Irritant

Formula: C₁₀H₇BrClN₃
MDL Number: MFCD11505046
CAS Number: 1208083-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR