2-Chloro-N-(4-bromophenyl)pyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

083975-1G
102841-210EA 467.69 USD
102841-210
2-Chloro-N-(4-bromophenyl)pyrimidin-4-amine ≥97%
2-Chloro-N-(4-bromophenyl)pyrimidin-4-amine

Hazard: Irritant

Formula: C₁₀H₇BrClN₃
MDL Number: MFCD09756924
CAS Number: 260046-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR