2-Chloro-N-(3-methoxyphenyl)pyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

083974-1G
102841-208EA 496.92 USD
102841-208
2-Chloro-N-(3-methoxyphenyl)pyrimidin-4-amine ≥97%
2-Chloro-N-(3-methoxyphenyl)pyrimidin-4-amine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃O
MDL Number: MFCD11043775
CAS Number: 191728-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR