2-Chloro-N-tetrahydro-2H-pyran-4-ylpyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

083971-1G
102841-202EA 609.23 USD
102841-202
2-Chloro-N-tetrahydro-2H-pyran-4-ylpyrimidin-4-amine ≥97%
2-Chloro-N-tetrahydro-2H-pyran-4-ylpyrimidin-4-amine

Hazard: Irritant

Formula: C₉H₁₂ClN₃O
MDL Number: MFCD09750123
CAS Number: 954229-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR