2-Chloro-4-(tetrahydro-2H-pyran-4-yloxy)pyrimidine ≥97%

Supplier: Matrix Scientific

083970-1G
102841-200EA 863.08 USD
102841-200
2-Chloro-4-(tetrahydro-2H-pyran-4-yloxy)pyrimidine ≥97%
2-Chloro-4-(tetrahydro-2H-pyran-4-yloxy)pyrimidine

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD11505043
CAS Number: 1216402-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR