2-Chloro-4-(cyclohexyloxy)pyrimidine ≥97%

Supplier: Matrix Scientific

083961-1G
102841-180EA 750.77 USD
102841-180
2-Chloro-4-(cyclohexyloxy)pyrimidine ≥97%
2-Chloro-4-(cyclohexyloxy)pyrimidine

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD09864957
CAS Number: 1086376-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR