2-Chloro-N-(3-fluorophenyl)pyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

083960-1G
102841-178EA 467.69 USD
102841-178
2-Chloro-N-(3-fluorophenyl)pyrimidin-4-amine ≥97%
2-Chloro-N-(3-fluorophenyl)pyrimidin-4-amine

Hazard: Irritant

Formula: C₁₀H₇ClFN₃
MDL Number: MFCD09864956
CAS Number: 898805-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR