4-(4-Iodo-1H-pyrazol-1-yl)piperidine ≥97%

Supplier: Matrix Scientific

083932-1G
102841-108EA 806.15 USD
102841-108
4-(4-Iodo-1H-pyrazol-1-yl)piperidine ≥97%
4-(4-Iodo-1H-pyrazol-1-yl)piperidine

Hazard: Irritant

Formula: C₈H₁₂IN₃
MDL Number: MFCD16622252
CAS Number: 1229457-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR