5-Chloro-1-ethyl-3-(trifluoromethyl)-1H-pyrazole ≥97%

Supplier: Matrix Scientific

083929-1G
102841-102EA 949.23 USD
102841-102
5-Chloro-1-ethyl-3-(trifluoromethyl)-1H-pyrazole ≥97%
5-Chloro-1-ethyl-3-(trifluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₆H₆ClF₃N₂
MDL Number: MFCD12026365
CAS Number: 665026-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR