2-(4-Bromo-1H-pyrazol-1-yl)-4-(trifluoromethyl)pyrimidine ≥97%

Supplier: Matrix Scientific

083923-1G
102841-086EA 693.85 USD
102841-086
2-(4-Bromo-1H-pyrazol-1-yl)-4-(trifluoromethyl)pyrimidine ≥97%
2-(4-Bromo-1H-pyrazol-1-yl)-4-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₈H₄BrF₃N₄
MDL Number: MFCD11505041
CAS Number: 1215458-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR